The Targeted Mass Exclusion filter removes the peaks of ions that you list in the mass list. You create a mass list by entering masses and charges in the Mass List table. You can also import the settings from (or export them to) a .cvs, a .txt, or an .xml file.
The following table describes the parameters in the Targeted Mass Exclusion Properties pane.
Parameter | Description |
|---|---|
Time Mode | Select from the following Time Mode options to enable scheduled or unscheduled targeted scans. For scheduled, the user can define the retention time and window size or the start and end time.
NOTE: This option is available only if the filter follows an allowable master scan. |
Include Intensity Threshold | When enabled, the target m/z intensity must be equal or greater than the specified intensity to exclude. Default: Deselected |
Add Mass List Targets Determined by Xcalibur AcquireX | (Available in Small Molecule application mode when you select "Enable Xcalibur AcquireX method modifications" from the Settings area.) Enable Xcalibur AcquireX Ab feature for this filter. AcquireX will add targets to the list. Default: Deselected |
Add Mass List Targets Determined by Xcalibur AcquireX Ab | (Available in Peptide application mode when you select "Enable Xcalibur AcquireX Ab method modifications" in the Settings area.) Enable Xcalibur AcquireX Ab feature for this filter. AcquireX will add targets to the list. Default: Deselected |
| |
Parameters in the Mass List Table | |
Compound | (Optional) Specify the name of the ion. |
Adduct | (Available when the Mass list Type is set to "m/z & z".) (Required when Formula is used) Define the adduct to be used for the defined compound formula or peptide sequence. |
m/z | (Available when the Mass List Type is set to "m/z" or "m/z & z".) Specify the m/z of the ion on which the MS performs the scan. When Formula is defined, the m/z is calculated automatically. The accepted m/z range will vary depending on analyzer used (Quadrupole or Ion Trap) and HMRn+ license activation. |
z | (Available when the Mass List Type is set to "m/z &z".) (Optional in some cases) Specify the expected charge state of the precursor ion, which is used to determine collision energy or reaction times (when applicable). When Formula is defined, specify the expected charge state of the precursor ion. This is used to calculate the m/z. Range: 0–24; default: 1 |
t start (min) | (Available when the Time Mode is set to Start/End Time.) The time (in minutes) to begin acquiring spectra for the defined target. |
t stop (min) | (Available when the Time Mode is set to Start/End Time.) The time (in minutes) to stop acquiring spectra for the defined target. |
RT Time (min) | (Available when the Time Mode is set to Retention Time Window.) The center of the Retention Time Window (in minutes). For example, when Retention Time is set to 10 and Window is set to 2, acquisition of spectra for the target will only occur between the 9 and 11 minutes. |
Window (min) | (Available when the Mass List Type is set to Retention Time Window.) The window width (in minutes), centered around the defined retention time. For example, when Retention Time is set to 10 and Window is set to 2, acquisition of spectra for the target will only occur between the 9 and 11 minutes. |
| |
Exclusion mass width | Defines how close a measured mass must be to the mass in the mass list to be considered the same mass.
|
Low | The low side of the mass tolerance width window. The low value defines how far below the reference mass the window starts on the low-mass side of the reference peak. Range (ppm): 0.01–1,000,000; default: 25 |
High | The high side of the mass tolerance width window. The high value defines how far above the reference mass the mass tolerance window extends on the high-mass side of the reference peak. Range (ppm): 0.01–1,000,000; default: 25 |
Exclude Isotopes | Excludes isotope peaks from consideration for further peak selection. Default: Selected |