When using a Targeted Loss Inclusion filter, if the system detects a targeted loss ion in the targeted loss mass list, and if all other conditions that are selected are satisfied, the filter triggers an MSn+1 scan on the targeted loss ion. You can also import the settings of the mass list table from (or export them to) a .cvs, a .txt, or an .xml file.
The following figure shows a flowchart of the Targeted Loss Inclusion filter.
The following table describes the parameters in the Targeted Loss Inclusion Properties pane.
Parameter | Description |
|---|---|
Mass List Type | Select from the following mass list types:
|
| |
Mass List Table | |
Compound | (Optional) Specify the name of the ion. |
m/z | (Available when the Mass List Type is set to "m/z".) Specify the m/z of the ion on which the MS performs the scan. When Formula is defined, the m/z is calculated automatically. The accepted m/z range will vary depending on analyzer used (Quadrupole or Ion Trap) and HMRn+ license activation. |
Formula | (Available when the Mass List Type is set to "M".) (Optional) Define the compound formula (for example, CH3OH) or peptide sequence (for example, MRFA). |
M | (Available when the Mass List Type is set to "M".) The targeted loss mass (in Daltons). Range: 0–100,000; default: 523.2577 |
z loss | (Available when the Mass List Type is set to "M".) Optional, in some cases. Specify the expected charge state of the precursor ion, which is used to determine collision energy or reaction times (when applicable). When 'Formula' is defined, specify the expected charge state of the precursor ion. This is used to calculate the m/z. Range: 0–24; default: 1 |
| |
Mass Tolerance | Defines how close a measured mass must be to the mass in the mass list to be considered the same mass.
|
Low | The low side of the mass tolerance width window. The low value defines how far below the reference mass the window starts on the low-mass side of the reference peak. Range (ppm): 0.01–1,000,000; default: 10 |
High | The high side of the mass tolerance width window. The high value defines how far above the reference mass the mass tolerance window extends on the high-mass side of the reference peak. Range (ppm): 0.01–1,000,000; default: 10 |
Trigger Only with Detection of at Least N Ions from the List | Trigger Only with Detection of at Least N Ions from the List. Default: Deselected When selected, the following option becomes available: n: Range: 1–1,000,000; default: 1 |
Only Ion(s) Within Top N Most Intense | Select to enable only ion(s) within Top N most intense. Default: Deselected When selected, the following option becomes available: n: Range: 1–1,000,000; default: 10 |
Only Ion(s) Above the Threshold (Relative Intensity, %) | Select to enable only ion(s) above the threshold (Relative Intensity, %). Default: Deselected When selected, the following option becomes available: Range: 1–100; default: 30 |
Trigger Only with Detection of Correct Charge State of the Product Ion | (Available when the Mass List Type is set to "M".) Performs subsequent scans only if the product ion charge state exactly matches the targeted ion charge state. Default: Selected |
Ignore Charge State Requirement for Unassigned Ions | (Available when the Mass List Type is set to "M".) When selected, ions that have an unassigned charge state do not need to meet the charge-state requirement in the table. Default: Selected |