For subsequent scans, the Targeted Mass Difference filter selects only ions that have one or two corresponding peaks with mass differences in the target mass list. There are several options that determine which ions to use for subsequent scans.
You create a target mass list by entering mass differences, not the m/z, in the Mass List table. You can also import the settings from (or export them to) a .cvs, a .txt, or an .xml file.
The following table describes the parameters in the Targeted Mass Difference Properties pane.
Parameter | Description |
|---|---|
Number of Precursors in the Targeted Group | Set to define mass difference requirements for precursor sets that contain 2 or 3 targets. Default: 2 |
| |
Parameters in the Mass List Table | |
Compound | (Optional) Specify the name of the ion. |
Formula | (Optional) Define the compound formula (for example, CH3OH) or peptide sequence (for example, MRFA). |
Delta M1 | Delta Mass between M1 and M2. Range: 0–2,000; Default: 8.01 |
Delta M2 | (Available when the "Number of precursors in the targeted group" is set to 3.) Set to define mass difference requirements for precursor sets that contain 2 or 3 targets. Range: 0–2,000; Default: 16.02 |
Partner Intensity Range Relative to the Most Intense Precursor (%) | The intensity ratio (as a percentage) of the partner ion signal intensity to the most intense precursor ion signal intensity that must be exceeded to trigger subsequent analysis. Range: 0–100; Default:10–100 |
Mass Tolerance | Defines how close a measured mass must be to the mass in the mass list to be considered the same mass.
|
Low | The low side of the mass tolerance width window. The low value defines how far below the reference mass the window starts on the low-mass side of the reference peak. Range (ppm): 0.01–1,000,000; default: 10 |
High | The high side of the mass tolerance width window. The high value defines how far above the reference mass the mass tolerance window extends on the high-mass side of the reference peak. Range (ppm): 0.01–1,000,000; default: 10 |
Perform subsequent scan on | For n = 2, the options are:
For n = 3, the options:
|
Charge state requirement | Select from the following options:
|