Use the Targeted Mass filter to select only the peaks of ions that are in the target mass list. You create a target mass list by entering masses and charges in the mass list table. You can also import the settings from (or export them to) a .cvs, a .txt, or an .xml file.
The following table describes the parameters in the Targeted Mass Properties pane.
Parameter | Description |
|---|---|
Mass List Type | Select from the following mass list types:
|
Time Mode | Select from the following Time Mode options to enable scheduled or unscheduled targeted scans. For scheduled, the user can define the retention time and window size or the start and end time.
NOTE: This option is available only if the filter follows an allowable master scan. |
Include Intensity Threshold | When enabled, the target m/z intensity must be equal or greater than the specified intensity to exclude. Default: Deselected |
Add Mass List Targets Determined by Xcalibur AcquireX | (Available when Enable Xcalibur AcquireX method modifications is selected from the Settings area.) Enable Xcalibur AcquireX feature for this method. Default: Deselected |
| |
Parameters in the Mass List Table | |
Compound | (Optional) Specify the name of the ion. |
Formula | (Available when the Mass List Type is set to "m/z & z".) (Optional) Define the compound formula (for example, CH3OH) or peptide sequence (for example, MRFA). |
Adduct | (Available when the Mass list Type is set to "m/z & z".) (Required when Formula is used) Define the adduct to be used for the defined compound formula or peptide sequence. |
m/z | (Available when the Mass List Type is set to "m/z" or "m/z & z".) Specify the m/z of the ion on which the MS performs the scan. When Formula is defined, the m/z is calculated automatically. The accepted m/z range will vary depending on analyzer used (Quadrupole or Ion Trap) and HMRn+ license activation. |
z | (Available when the Mass List Type is set to "m/z &z".) (Optional in some cases) Specify the expected charge state of the precursor ion, which is used to determine collision energy or reaction times (when applicable). When Formula is defined, specify the expected charge state of the precursor ion. This is used to calculate the m/z. Range: 0–24; default: 1 |
t start (min) | (Available when the Time Mode is set to Start/End Time.) The time (in minutes) to begin acquiring spectra for the defined target. |
t stop (min) | (Available when the Time Mode is set to Start/End Time.) The time (in minutes) to stop acquiring spectra for the defined target. |
RT Time (min) | (Available when the Time Mode is set to Retention Time Window.) The center of the Retention Time Window (in minutes). For example, when Retention Time is set to 10 and Window is set to 2, acquisition of spectra for the target will only occur between the 9 and 11 minutes. |
Window (min) | (Available when the Mass List Type is set to Retention Time Window.) The window width (in minutes), centered around the defined retention time. For example, when Retention Time is set to 10 and Window is set to 2, acquisition of spectra for the target will only occur between the 9 and 11 minutes. |
| |
Mass Tolerance | Defines how close a measured mass must be to the mass in the mass list to be considered the same mass.
|
Low | The low side of the mass tolerance width window. The low value defines how far below the reference mass the window starts on the low-mass side of the reference peak. Range (ppm): 0.01–1,000,000; default: 25 |
High | The high side of the mass tolerance width window. The high value defines how far above the reference mass the mass tolerance window extends on the high-mass side of the reference peak. Range (ppm): 0.01–1,000,000; default: 25 |
Ignore Charge State Requirement for Unassigned Ions | (Available when the Mass List Type is set to "m/z & z".) When selected, ions that have an unassigned charge state do not need to meet the charge-state requirement in the table. Default: Selected |
Set Collision Energy per Compound | When selected, the HCD Collision Energy column is added to the table. NOTE: When Xcalibur AcquireX is enabled, Collision Energy cannot be set for individual targets. |
Perform dependent scan on most intense ions if no targets are found | When selected, if insufficient number of targets on the inclusion list are detected for the scan cycle, the system will perform dependent scans based on intensity. Default: Deselected |
Use Group IDs | When checked, the elements in the list can be grouped by a unique identifier, 'Group ID'. Groups are subsets of the whole list. When determining the inclusion conditions, groups are considered independently of each other. Default: Deselected |