Use the Precursor Selection Range filter to select precursors from a narrower range than the range that was scanned. You can set a mass-to-charge ratio selection range (m/z). After an MSn scan, you can set a range that is relative to the precursor ion (%). You can add a precursor selection range filter after any scan.
NOTE
You cannot set the precursor selection range outside of the scan range.
The following table describes the parameters in the Precursor Selection Range properties pane.
Parameter | Description |
|---|---|
Selection Range Mode | Select to define a mass range or have it automatically set relative to the precursor m/z.
|
Mass Range (m/z) | Define mass range from which precursors will be selected for data-dependent scans. This filter is optional. If the filter is not used, the mass range defaults to the scan range of the master scan. Use this filter to define a subset of the scan range from which precursors will be selected for data-dependent scans. |
Range Relative to Parent m/z (%) | Define mass range (relative to parent) from which precursors will be selected for data-dependent scans. This filter is optional. If the filter is not used, the mass range defaults to the scan range of the master scan. Use this filter to define a subset of the scan range from which precursors will be selected for data-dependent scans. In this example, if the precursor is at m/z 500 and the range relative to parent m/z is set to 10–150%, the selection range for subsequent scans will be from m/z 50–750. |