The minimum hardware and software configurations required for Orbitrap Exploris Series 4.4 Instrument Control Software operation are as follows:
System | Requirements |
|---|---|
PC | 3.0 GHz Quad Core Intel™ Processor 32 GB RAM 512 GB SSD Hard Drive DVD-ROM Drive Display Monitor Resolution of 1920 × 1080 Two Network Interface Cards (NIC), 1000 MBit/s |
Software | Microsoft™ Windows™ 10 Enterprise 2016 LTSB, 2019 LTSC or 2021 LTSC Thermo Scientific™ Xcalibur™ 4.7 |
Note that the Orbitrap Exploris Series 4.4 Instrument Control Software was only tested within the delivered composition.