The proper performance of your system can be affected by the waste and exhaust arrangements for the instrument. The vacuum waste and the solvent waste must be vented separately, and the wastes must be collected and disposed of properly.
IMPORTANT
You are responsible for providing proper waste and exhaust systems for the operation of your system.
Contents