Place the outlets for the MS system, forepump, and LC system behind the LC system's workbench. Place the outlets for the data system—computer, monitor, Ethernet switch, and (optional) printer—behind the data system workbench. For the LC system, use wall outlets. Additional power outlets might be required for testing and cleaning equipment, such as an oscilloscope and ultrasonic bath.
The maximum load for a 120 Vac fourplex outlet is typically 20 A, and the maximum load for a 230 Vac fourplex outlet is typically 16 A. Thermo Fisher Scientific recommends that you have at least 6 spare outlets behind the system and 3 spare outlets close to the workbench space within your laboratory.