Thermo Fisher Scientific provides six power cords for the mass spectrometer, forepump, data system, monitor, and Ethernet switch. They are approximately 2.5 m (8 ft) long. One power cord fits into a standard IEC 60320 C19 socket on the forepump. The other cords fit into standard IEC 60320 C13 sockets on the mass spectrometer and the other system components.
TIP
The MS provides the electric power for the syringe pump. The MS can also supply the electric power for the Ethernet switch. This allows positioning the Ethernet switch near the MS and the LC.
NOTICE
The instrument is equipped with a detachable mains cable. Use only mains supply cords with an adequate rating. All spare parts and consumable items used must be approved by Thermo Fisher Scientific.
Depending on the location of your laboratory, various types of plugs may be required. The table gives an overview of the possible wall outlets. Write the part number of the corresponding country kit into the Installation Request Form at the beginning of this guide and send it to the factory before the installation.
NOTICE
For more information on the correct cable kit, contact your certified Thermo Fisher Scientific service representative.
P/N | Designation | Socket for source vacuum pump | Socket for MS system, PC, monitor, and Ethernet switch | Qty. |
|---|---|---|---|---|
BRE0016334 | Country Kit Continental Europe (Afghanistan, Algeria, Angola, Armenia, Austria, Azores [Portugal], Belarus, Belgium, Benin, Bosnia-Herzegovina, Bulgaria, Burundi, Cambodia, Cameroon, Canary Islands [Spain], Rep. of Cape Verde, Central African Rep., Chad, Chile, Congo, Croatia, Czech. Rep., Rep. of Djibouti, Egypt, Equatorial Guinea, Estonia, Ethiopia, Finland, France, French Guiana, Gabon, Germany, Greece, Guadeloupe, Guinea, Guinea-Bissau, Hungary, Iceland, Indonesia, Iran, Iraq, Ivory Coast, Jordan, Kyrgyzstan, Laos, Latvia, Lebanon, Liberia, Libya, Lithuania, Luxembourg, Madagascar, Mali, Martinique [France], Mauritania, Moldova, Monaco, Mongolia, Morocco, Mozambique, Netherlands, New Caledonia, Niger, North Korea, Norway, Oman, Paraguay, Peru, Poland, Portugal, Romania, Russia Federation, Rwanda, Saudi Arabia, Senegal, Slovakia, Slovenia, South Korea, Spain, Svalbard [Norway], Sweden, Syria, Thailand, Togo, Tunisia, Turkey, Ukraine, Uruguay, Vietnam, and Yugoslavia) | CEE 7 plug | CEE 7 plug | 1+5 |
BRE0017109 | Country Kit Australia/New Zealand (American Samoa, Australia, Christmas Is. [Austral.], Cocos Is. [Austral.], Cook Is. [N.Z.], New Zealand, Papua New Guinea, Fiji, Tonga, and Western Samoa) | AS 3112 plug | AS 3112 plug | 1+5 |
BRE0017112 | Country Kit United Kingdom/Ireland (Anguilla [U.K.], Antigua, Bahrain, Bangladesh, Botswana, Burkina Faso, Channel Islands, Cyprus, Dominica, England, Gambia, Ghana, Gibraltar, Grenada, Hong Kong, Ireland, Rep. of Kenya, Kuwait, Lesotho, Macao, Malawi, Malaysia, Maldives, Malta, Mauritius, Myanmar [formerly Burma], Rep. of Namibia [W.S.Africa], Nigeria, North Ireland, Qatar, Scotland, Seychelles, Sierra Leone, Singapore, South Africa, Sri Lanka, St. Kitts & Nevis, St. Lucia, St. Vincent, Sudan, Swaziland, Tanzania, Uganda, United Arab Emir., United Kingdom & Ire., Wales, Yemen, Zambia, and Zimbabwe) | BS 1363 plug | BS 1363 plug | 1+5 |
BRE0017114 | Country Kit Denmark (Denmark and Greenland [Den.]) | DS 60884-2-D1 plug | DS 60884-2-D1 plug | 1+5 |
BRE0017116 | Country Kit France/Belgium (Belgium and France) | CEE 7 plug | CEE 7/7 plug | 1+5 |
BRE0017124 | Country Kit India/ South Africa (India, Nepal, Pakistan, Pitcairn Is. [U.K.], and Somalia) | BS 546 15 A | BS 546 15 A | 1+5 |
BRE0017127 | Country Kit Israel (Israel) | SI 32 plug | SI 32 plug | 1+5 |
BRE0017129 | Country Kit Italy (Italy) | IT2-16P | YP-45 | 1+5 |
CEE 7 plug | CEE 7 plug | 1+1 | ||
BRE0017132 | Country Kit Japan (Japan) | NEMA L6-20P | JIS C 8303 | 1+4 |
NEMA L6-20 | 2 | |||
BRE0017135 | Country Kit NA (Aruba, Bahamas, Barbados, Belize, Bermuda, Bolivia, Brazil, Canada, Cayman Islands, Colombia, Costa Rica, Cuba, Curacao Is., Dominican Rep., Ecuador, El Salvador, Guam, Guatemala, Guyana, Haiti, Honduras, Jamaica, Mexico, Montserrat, Neth. Antilles, Nicaragua, Panama, Philippines, Puerto Rico, St. Pierre & Miquelon [Fr.], Suriname, Tahiti, Taiwan, Thailand, Trinidad & Tobago, United States, Venezuela, and Virgin Islands) * One socket supplies the source vacuum pump, and the other socket supplies the mass spectrometer. | NEMA L6-20* | NEMA 5-15 | 1+4 |
NEMA L6-20* | 1 | |||
BRE0017141 | Country Kit Switzerland (Liechtenstein and Switzerland) | SW2-16P | SEV 1011: 2009 | 1+5 |
BRE0017143 | Country Kit Brazil (Brazil) | NBR 14136 (BR3-20P) | NBR 14136 (BR3-20P) | 1+5 |
BRE0017145 | Country Kit Argentina (Argentina) | IRAM2073 AR2-20P | IRAM2073 | 1+5 |
BRE0017147 | Country Kit China (China) | CH2-16P | PRC/3 | 1+5 |
Power cables and connectors for the options are standard equipment delivered by the manufacturers.