The instrument can withstand electromagnetic fields of 1 V/m in the frequency range 26 MHz to 1 GHz without any influence to operation.
The instrument is designed to work in a controlled electromagnetic environment. Do not use radio frequency transmitters, such as mobile phones, in close proximity to the instrument.
If strong radio transmitters are operating close to your laboratory, you should contact your local Thermo Fisher Scientific representative for advice. Because of the complexity of such influences, no general suggestion can be given in this guide.