The instrument is designed to be operated in an environment that is rated with pollution degree 2. This means that only non-conductive pollution occurs.
The relative humidity of the operating environment must be between 20 and 80%, with no condensation. Thermo Fisher Scientific recommends to install a temperature/humidity monitor in your laboratory. This makes sure that the laboratory is always within the specifications for temperature and humidity.
NOTICE
Operating the instrument at very low humidity might cause the accumulation and discharge of static electricity, which can shorten the life of electronic components. Operating the system at high humidity might cause condensation, oxidation, and short circuits, and also blocks the filters on the cooling fans.