The body of the Orbitrap Excedion Series MS has maximum dimensions of h 703 mm (27 11⁄16 in.), w 535 mm (21 1⁄16 in.), l 999 mm (39 5⁄16 in.). The figure shows a schematic view of the instrument with important instrument dimensions.
The body of the Orbitrap Excedion Series MS has maximum dimensions of h 703 mm (27 11⁄16 in.), w 535 mm (21 1⁄16 in.), l 999 mm (39 5⁄16 in.). The figure shows a schematic view of the instrument with important instrument dimensions.