The LC/MS system is designed to be placed on top of a workbench with its rear panel against a wall. To set up a typical LC/MS system, Thermo Fisher Scientific recommends that you have a minimum of two workbenches.
Table Minimum workbench surface dimensions
Equipment
Surface
Data system (Computer workstation)
120 × 100 cm (47 ¼ × 39 3⁄8 in.)
LC/MS system
150 × 100 cm (59 1⁄16 × 39 3⁄8 in.)
The workbench for the LC/MS system must be capable of supporting the weight of the mass spectrometer (about 110 kg) plus the weight of any option (liquid chromatograph, for example) and stand in a secure and level position.
IMPORTANT
Only workbenches with four legs provide sufficient stability for the instrument and other equipment. The workbench top must be dry and clean (free of grease). Thermo Fisher Scientific recommends that you use a workbench with a skidproof top.
Clearance
Follow these clearance guidelines for the workbenches:
  • Place the data system workbench and the LC/MS workbench adjacent to each other to prevent strain on the interconnecting Ethernet communications cables.
  • Make sure that you have the following minimum clearances:
    • 900 mm (35 ½ in.) between the top of the system and any shelves above it.
    • 400 mm (16 in.) between the left side or the right side of the instrument and any other components.
Spacers at the rear side of the instrument provide sufficient space for airflow.
To allow shutting off the mass spectrometer in an emergency, free access to the power column at the right side of the instrument must be possible at any time.
NOTICE
Do not block the ventilation slots of the instrument. Items might fall behind the instrument and inhibit airflow, which can cause the system to overheat.