The table summarizes the most important technical data of the Orbitrap Excedion Series systems. See the respective chapters of the manual for details and additional instrument properties.
Table Technical data
Parameter
Specification
Value
Instrument Properties
Mass Spectrometer
Length × width × height
763 × 534 × 703 mm
Weight
110 kg
Forepump1)
Length × width × height
704 × 308 × 344 mm
Weight
78 kg
Complete system
(incl. forepump and data system)
Noise emission
Below 70 dB(A)
Heat generation
2000 W
Power Requirements
Mass Spectrometer
Input
Nominal voltage
208–240 V AC, 50/60 Hz
Power
apparent power: 800 VA; effective power: 750 W
Fuse2)
15/16 A (tripping characteristic C)
Output
208–240 V AC, 50/60 Hz, 3 A total
Protection class
Class I
Overvoltage category
II
Forepump
Input
Nominal voltage
200–230 V AC, 50/60 Hz
Power
700 W
Fuse2)
15/16 A (tripping characteristic C)
Data System
Input
Nominal voltage
100–240 V AC, 50/60 Hz
Fuse
15/16 A
Gas Requirements
Source gas (minimum requirement)
Type
Nitrogen
Purity
99% or better (high purity)
Supply rate
max. 45 NL/min
Pressure
0.6±0.05 MPa
HCD gas (mandatory)
Type
Nitrogen
Purity
99.999% or better (ultra high purity)
Supply rate 3)
max. 0.03 LN/min
Pressure
0.6±0.05 MPa
Operating Environment
Laboratory temperature
18–27 °C
Max. temperature fluctuation
0.5 °C/10 min
Humidity
20–80%, non-condensing and non-corrosive atmosphere
Pollution degree
2
Max. altitude
3000 m above sea level
Pump exhaust requirements
Inrush flow rate
10 L/min
Continuous flow rate
7 L/min
Ion source exhaust requirements
Maximum flow rate
45 L/min
TIP
The unit NL refers to liters at normal conditions, measured at a temperature of 0 °C and a pressure of 1013.25 hPa. These reference conditions are indicated by the added letter “N” in the unit of volume used.