The mass spectrometer requires a syringe pump for delivering sample solution from a syringe into the API ion source. A suitable syringe pump is shipped with the mass spectrometer. Place the syringe pump next to the API source in the accessories cabinet of the mass spectrometer.