If your local area is susceptible to corrupted power or power disruptions, install a UPS in your laboratory.
TIP
For compliance and safety, make sure that your UPS devices are certified by recognized domestic and international organizations (for example, UL, CSA, TÜV, and VDE).