The table below lists all known issues that exist in the Orbitrap Astral Series 2.0 SP1 Instrument Control Software release.
ID | Severity | Title |
|---|---|---|
347288 | 4 - Low | Tune: Setting incorrect Syringe Pump flow rate displays wrong message “the syringe may be out of sample”. Remedy: Remove and place the syringe again into the pump to release the backpressure. |
357067 | 3 - Medium | Isolation m/z range below m/z 40 does not trigger a validation error when resulting from an inappropriate isolation offset setting. Remedy: Enter an isolation offset value that complies with accepted isolation range. |
387627 | 4 - Low | RF values are not converted to account for differences between S-lens and funnel when importing OE120/240/MX methods. Remedy: Please manually adjust the RF value accordingly. |
404844 | 3 - Medium | Xcalibur: If the file name contains the symbol μ, two raw data files are generated instead of one. Remedy: Do not use symbol μ in file name. |
614321 | 3 - Medium | Ion Foil Correction and Astral Mass Stabilization can fail due to uncalibrated ICS source. Remedy: Perform System Calibration. |
676639 | 3 - Medium | FAIMS: Temperature differences above 20 °C between inner and outer electrode might not be reachable and cause timeouts. Remedy: When using "User Defined" FAIMS Mode in Method Editor, adjust FAIMS Inner Electrode Temp/FAIMS Outer Electrode Temp in order keep the temperature difference between the two electrodes below 20 °C. |
813657 | 3 - Medium | Acquisition time not properly reported in rawfile with tMS2-only method not starting acquisition at t=0 min. Remedy: Accept the incorrect report of acquisition time in rawfile (not affecting acquisition) or modify the method so that actual acquisition is started at t=0 min (e.g., by including a full scan experiment starting at t=0 min). |
820630 | 3 - Medium | UI: ME: tSIM and tMS2: setting maxIT and microscans in table/scan property. Remedy: Revert "Maximum Injection Time" or "Microscan" as a scan property. |
824228 | 3 - Medium | DDA performed in small molecule application mode and with m/z of selected precursor ion varying substantially may result in unstable TIC for full scan MS and ddMS2 overfilling (for high mass precursor ion). Remedy: Restrict the full scan mass range by using, ideally, a last m/z value not exceeding five times the first m/z value. |
827732 | 2 - High | UI: ME: TMT HR Mode available in Small Molecule and Intact Protein Modes. Remedy: Do not use TMT HR Mode in "Small Molecule" and "Intact Protein" modes as it is not relevant. |
814458 | 3 - Medium | Scan Range mode set to "Auto-Extended" in some of the scans of a formerly saved method (using small molecule application mode) is changed to "Define m/z Range" when re-opened. Remedy: The method works as expected at the time of saving. If the method is re-opened for modification, make sure Scan Range mode is set again to "Auto-Extended" before saving the method. |
829920 | 3 - Medium | Methods including eDR m/z windows having first mass higher than m/z 2500 with "eDR Window Type" set to "Auto" do not report error and can be saved without warning, but do not pass method validation when executing a sequence in Xcalibur. Remedy: Set "eDR Window Type" to "User Defined" before saving the method to confirm m/z window values are in accepted range. |
830065 | 3 - Medium | Raw data files acquired using DIA methods with ''DIA Window Mode'' set to "Center Mass" report DIA m/z windows table with headers "m/z" and "Q1 Resolution (m/z)" instead of "Center Mass (m/z)" and "Window Width (m/z)", respectively, in summary section. |
845079 | 3 - Medium | Service Diagnostic Tool running in the background can cause interruption in sequence execution Remedy: Close Service Diagnostic Tool after service and troubleshooting tasks. |