The table below lists defect fixes in the Orbitrap Astral Series 2.0 SP1 Instrument Control Software release that were implemented since the Orbitrap Astral Series 2.0 Instrument Control Software release.
Table Changes realized with Orbitrap Astral Series 2.0 SP1 Instrument Control Software relative to Orbitrap Astral Series 2.0 Instrument Control Software
ID
Severity
Title
Defect fixes
823734
3 - Medium
Spray stability evaluation fails although spray was stable before start of the evaluation
834056
3 - Medium
SureQuant and Hybrid DIA methods may not work properly with certain Peripheral settings
820722
2 - High
Sequence execution interrupted due to MS response timeout.
Note: The fix reduces the frequency of sequence interruption issue but does not fully address it. It is also important to make sure that the remedy of Known Issue 845079 is applied.
The table below lists new features, improvements and defect fixes in the Orbitrap Astral Series 2.0 Instrument Control Software release that were implemented since the Orbitrap Astral Series 1.1 Instrument Control Software release.
Table Changes realized with Orbitrap Astral Series 2.0 Instrument Control Software relative to Orbitrap Astral Series 1.1 Instrument Control Software
ID
Severity
Title
New features
488673
—
Orbitrap Astral Zoom: Faster Ion Optics Switching: Higher acquisition rate with improved ion optics switching times
740686
—
Orbitrap Astral Zoom: Faster Ion Processor Timings: Higher acquisition rate due to faster ion transfer in the Ion Processor
491850
—
Orbitrap Astral Zoom: Pre-Accumulation: The user needs bent-flatapole pre-accumulation
595718
—
Orbitrap Astral Zoom: Application Mode: The user needs a "Low Input" Application Mode
595715
—
Orbitrap Astral Zoom: TMT 32-Plex: The user needs a TMT HR mode to resolve the signal of all reporter ions
449007
—
Orbitrap Astral Zoom: Peak Picking: The user needs Full Astral MS2 Spectrum Peak Picking
491831
—
Orbitrap Astral Zoom: SureQuant Hybrid DIA: The user needs to perform SureQuant type and derived Hybrid DIA acquisition using Astral analyzer
747451
—
Orbitrap Astral Zoom: Faster Stepped NCE in IRM: The user needs faster Astral MS2 acquisition when using stepped Collision Energy
424340
—
Orbitrap Astral Zoom: The user needs Top-Down Acquisition with Astral MS2 scans
169738
—
Orbitrap Astral Zoom: eDR: The user needs to perform eDR Full Scan-type ion detection
650535
—
Orbitrap Astral Zoom: tSIM msx: The user needs the ability to execute methods with multiplex tSIM with Orbitrap analyzer
Improvements
745468
—
Astral MS2 Scan Range: The user needs expanded functionalities of Astral MS2 Scan Range
745777
—
System Calibrations must provide improved user experience
749850
—
Add "Exclude isotopic cluster" option to Targeted Mass Exclusion filter
749864
—
The user needs individual maxIT settings in DDA w/ inclusion list (directed-ddA)
778704
—
RAW File Scan Header: The user needs the ability to differentiate tMS2 and DIA scans via the scan header
Defect fixes
639247
3 - Medium
UI: ME: Orbitrap Astral Method Summary for DIA does not report the correct parameter used for Loop Control
747865
3 - Medium
UI: ME: Factor 15 Rule is applied for Astral MS2 scan range unexpectedly
784006
3 - Medium
UI: ME: RAW File Instrument Method erroneously reports Astral Resolution
808914
3 - Medium
UI: XML Method Modification has dysfunctions
639762
3 - Medium
After changing at setting in Tune, while the Instrument is in Intact Protein mode, Injection Time is unregulated
631487
3 - Medium
Method Execution: nCE values are translated in CE values above 200 V
763179
1 - Critical
RunStart ICS degrades mass accuracy
776401
1 - Critical
Automatic Ion Foil Offset Adjustment leads to failing mass evaluations with large mass errors
779710
4 - Low
Tune: Calibration and Diagnostics Start button - procedure "cancelled by user", after internal error
The table below lists new features, improvements and defect fixes in the Orbitrap Astral Series 1.1 Instrument Control Software release that were implemented since the Orbitrap Astral Series 1.0 SP3 Instrument Control Software release.
Table Changes realized with Orbitrap Astral Series 1.1 Instrument Control Software relative to Orbitrap Astral Series 1.0 SP3 Instrument Control Software
ID
Severity
Title
New features
491821
—
Stepped CE Astral: The user needs the ability to perform the acquisition of single MS2 scan with multiple Collision Energy values when using Astral detector Type
425845
—
Full Profile OT: The user needs the ability to perform Orbitrap MS1 and MS2 acquisition with Full Profile in Intact Protein Mode
Improvements
610911
—
NSI: The user needs an updated default value of Positive Spray Voltage for NSI in ME and Tune
431751
—
Orbitrap Astral shall automatically correct the ion foil voltage using the ICS
597081
—
FAIMS CV switching performance shall be improved
496128
—
UI: ME: The user needs the "Lock mass injection" to be a parameter in the scan properties for each scan type
407868
—
Method Editor: The System Templates must be saved in "Astral" folder
434211
—
UI: ME: The user needs the possibility to choose "Use EASY-IC" On/Off for each experiment/scan type in Timed Mode
469200
—
UI: ME: The user needs to be able to see the displayed absolute AGC target values
408025
—
Tune: Favorites/UserSettings and History information must be saved in "Astral" folders
448670
—
The user needs the ability to monitor pressure readbacks for Astral Analyzer in Tune
Defect fixes
458725
3 - Medium
MongoDB uses large memory (>3GB) at background
459860
3 - Medium
If in DIA experiments (using Astral detector) a Loop Time is defined, as shorter than the Orbitrap transient time set in another experiment of the method, the instrument schedules not enough DIA scans to match the expected loop control time
594611
3 - Medium
Fluxscans are not used in edge cases (fm/lm +/- isolation width)
597496
3 - Medium
DIA: Methods miswriting mass window centres as 524.265 in raw file instrument method
598230
3 - Medium
Missing Plot: Mass Calibration
591844
3 - Medium
Tune: Pressure Mode always set to Standard Pressure after startup
592082
3 - Medium
Tune UI: Astral Turbo pumps readbacks are swapped
467823
3 - Medium
Issue with DIA Windows Definition with Windows Optimization "On" under certain conditions
470082
3 - Medium
Max IT information is not copied when using copy/paste functionality of MS2 information - instead default max IT (10 ms) is used
439849
3 - Medium
"Use EASY-IC" dropdown is not available if LM correction mode is set to "timed"
412392
2 - High
Instrument reported to be set to standby because of open source, but continued the sequence
424847
3 - Medium
The search window for the EASY-IC lockmass is shifted by outliers
426171
3 - Medium
"Bakeout cooling time" and other ICB/HICB timers are reset by software update
415413
3 - Medium
Astral Mass Stabilization is performed at different timepoints within a LC-MS run
455566
3 - Medium
Syringe does not get turned off during Electronics Smart Calibration in User System Calibration
456040
2 - High
Setting the Funnel RF Level to default is not in Astral Default Scan Settings
The table below lists defect fixes in the Orbitrap Astral Series 1.0 SP3 Instrument Control Software release that were implemented since the Orbitrap Astral Series 1.0 SP2 Instrument Control Software release.
Table Changes realized with Orbitrap Astral Series 1.0 SP3 Instrument Control Software relative to Orbitrap Astral Series 1.0 SP2 Instrument Control Software
Defect Fixes
ID
Severity
Title
466217
1 - Critical
Large Time Gap between MS1 and data-dependent MS2 scan.
553186
2 - High
Insufficient IRM settling time causing unstable fragmentation pattern in Orbitrap MS2 scans.
452322
1 - Critical
Ion Control not working well under low intensity, high background conditions.
414707
3 - Medium
False positive initial bus error warnings from DPRTrap Board.
427472
2 - High
Detector Calibration failing with “Too few ions in focused peak.”
449905
2 - High
Suboptimal setting of channel saturation threshold.
234095
2 - High
Astral analyzer pressure readbacks not displayed in Tune.
The table below lists defect fixes in the Orbitrap Astral Series 1.0 SP2 Instrument Control Software release that were implemented since the Orbitrap Astral Series 1.0 SP1 Instrument Control Software release.
Table Changes realized with Orbitrap Astral Series 1.0 SP2 Instrument Control Software relative to Orbitrap Astral Series 1.0 SP1 Instrument Control Software
Defect Fixes
ID
Severity
Title
357681
4 - Low
Method execution waits for temperatures to stabilize while 'Wait for Temperatures to stabilize' option is unchecked in Instrument Configuration.
361874
3 - Medium
Missing fields in the scan header/trailer.
398882
3 - Medium
AGC calculations for method using more than one FAIMS CV (delta>1) are based on single CV.
403091
3 - Medium
Raw file: Scan headers of ASTMS scans report irrelevant FT resolution.
407881
3 - Medium
Data Acquisition using lock mass correction is not aborted while Internal Calibrant Source is faulty/defective.
396718
1 - Critical
Data Acquisition: FAIMS occasionally returns “source is open” right after the start of a method while source is closed.
416835
1 - Critical
Both the restart from an error as well as quick system restart fail with a restart loop.
418457
1 - Critical
Orbitrap mode scan rate too low after reboot of system.
424800
3 - Medium
Orbitrap easyIC lockmass correction (timed / Scan-to-Scan modes) is not functional if the first mass of the scan range is above m/z 275.
424984
1 - Critical
Failing Signal Stability Evaluation - high TIC variations in Orbitrap signal.
425564
3 - Medium
Bake-out plot is not redisplayed after the system entered standby.
427904
3 - Medium
Block voltage insufficient for Curve Corrector on systems with large non-zero curve corrector settings.
428554
3 - Medium
Activating microscans shifts the mass centroid in Astral spectra.
429956
3 - Medium
TNG System Check Negative: system
429981
3 - Medium
“Short burn-in of ICS” is always run during System Check.
430132
3 - Medium
Previous plot is not redisplayed after Astral analyzer polarity switch.
430259
1 - Critical
Discharge evaluation not actually setting to negative mode during discharge event.
431767
3 - Medium
Missing Quick Gas Capillary Check to allow the user to assess if a capillary is blocked.
433742
3 - Medium
Scan Start Timing fallback option may lead to out of order retention times.
434485
3 - Medium
Missing checks in Tune Diagnostics.
436967
3 - Medium
Astral service bundle mostly empty (no astral data).
442099
2 - High
System Calibration: Quad RF DDS frequency does not get set always on boot.
450428
1 - Critical
After performing Mass and System check from Tune, a drift of about 1 m/z unit may be observed in mass spectra acquired with Astral detector.
The table below lists defect fixes in the Orbitrap Astral Series 1.0 SP1 Instrument Control Software release that were implemented since the Orbitrap Astral Series 1.0 Instrument Control Software release.
Table Changes realized with Orbitrap Astral Series 1.0 SP1 Instrument Control Software relative to Orbitrap Astral Series 1.0 Instrument Control Software
Defect Fixes
ID
Severity
Title
412822
1 - Critical
Occasionally spikes can be observed in ion chromatogram while ion spray stable.
419732
1 - Critical
EASY-IC: Lockmass expected position does not change with polarity.
421872
1 - Critical
XML method modification interface is not functional.
419661
2 - High
System Calibration/Check: Change PrOSA Scaling specifications.
416053
3 - Medium
System Calibration/Check: HCD Fragmentation and Trapping Efficiency Check leads to a SW internal failure (occasionally).