The instruments listed in the table below are supplied and supported in this release.
Instrument Control Software Version | Version No. | Instrument | |
|---|---|---|---|
Orbitrap Astral | Orbitrap Astral Zoom | ||
1.0 | 1.0.100.11 | ü | – |
1.0 SP1 | 1.0.100.14 | ü | – |
1.0 SP2 | 1.0.100.28 | ü | – |
1.0 SP3 | 1.0.100.40 | ü | – |
1.1 | 1.1.477.46 | ü | – |
2.0 | 2.0.690.11 | ü | ü |
2.0 SP1 | 2.0.690.13 | ü | ü |