Procedure
- Place the quadrupole straight down onto the holders.
NOTICE
Do not to bump the ends of the quadrupole on any adjacent parts.
- Make sure that the quadrupole seating is correct. Check the copper clips on the handle for a proper seating to make sure that they will not fall into the chamber.
- Tighten the four 3 mm hex screws.
Re-installing the quadrupole - Close the analyzer chamber.
- Make sure that there are no particles on the black sealing of the chamber lid. If necessary, remove them with a clean lint-free tissue.
- Use the handle to close the chamber lid.