Clean the bent flatapole
Procedure
- Purge the component from any particles with a strong stream of oil-free nitrogen gas.
Clean the bent flatapole exit lens
Procedure
- Clean the inside of the orifice with a conical swab that is soaked with a 50:50 isopropanol/water solution (LC/MS grade).
Using a swab - Clean the neutral spot with a lint-free wipe that is soaked in a 50:50 isopropanol/water solution. The spot is on the right side of one of the metal cylinders.
- Clean the lens orifice from both sides with a lint-free wipe that is soaked in a 50:50 isopropanol/water solution.
- Clean the lens orifice from both sides with a lint-free wipe that is soaked in 100% isopropanol.
- Dry the component with a strong stream of oil-free nitrogen gas to make sure that the solvent evaporates completely.