When you do extended maintenance on the Orbitrap Astral system, pay attention to the safety guidelines that follow. Also refer to the chapter “Safety” of the Orbitrap Astral Operating Manual for comprehensive information about machine safety.
WARNING
High Voltage
Hazardous electric voltage that can cause an electric shock is used in the instrument. To make sure that the instrument is free from all electric current, always disconnect the power cords of the mass spectrometer and the forepump before you try any type of maintenance.
CAUTION
Crushing Hazard
The hinged lid of the analyzer chamber might unexpectedly fall down when it is not in its resting position. Make sure that the lid is secure when you do work on the components inside.
NOTE
Avoid exposure to potentially harmful materials.
Read the SDS for each chemical you use. Store and handle all chemicals in accordance with standard safety procedures. Always wear protective gloves and safety glasses when you use solvents or corrosives. Also, contain waste streams, use proper ventilation, and dispose of all laboratory reagents according to the directions in the SDS.
NOTICE
- When you do work within the system, you should wear clean, powder-free and lint-free gloves. When you handle solvents and chemicals, the glove material should at least provide temporary resistance to the solvents. Usage of nitrile gloves is recommended.
- Dirty tools can contaminate your system. Keep the tools clean and use them exclusively for the maintenance of the ion optics.
- Store the disassembled parts on a clean, lint-free surface (for example, aluminum foil). Store the already cleaned optics under a closed hood or place them into the instrument as soon as possible and close the lid.