This document is an addendum to the Orbitrap Astral Operating Manual. It describes the maintenance for ion optic components that are accessible after removing the top cover of the instrument. This document is designated for customers who have received a special training about extended maintenance for the quadrupole and the bent flatapole.
Component | Frequency | See |
|---|---|---|
Quadrupole | Clean if quadrupole performance evaluation indicates contamination. | |
Bent flatapole | When you clean the quadrupole. |