Run installation diagnostics
After you attach the OptiSpray ion source to the mass spectrometer and test the camera views, use the source diagnostic cartridge with the Tune application to perform the following installation diagnostics:
-
Hardware readbacks: Verifies that the OptiSpray ion source is properly connected to the MS system and that all internal electrical boards are functioning.
-
Motor movement: Verifies that the motors move smoothly and reach their set positions.
-
Cartridge heater: Verifies that the cartridge heater is functioning properly and can reach its set temperature.
-
Cartridge sheath gas: Verifies that sheath gas is being delivered.
TIP
For a video demonstration of this process, see the following video.
Prerequisites
-
OptiSpray ion source, sweep cone, and source diagnostic cartridge are installed.
Procedure
-
Set the cartridge temperature to 50 °C:
- On the Ion Source page of the Tune application, select the Ion Source tab.
-
Set the Cartridge Temp to 50 °C, then select Apply.
Figure 1. Set the cartridge temperature The Status icon turns green when the cartridge reaches the set temperature (within 5 minutes).
-
Ensure the cartridge is in the inactive position:
- From the Tune ribbon, select the cartridge status drop-down menu.
-
If not already set to Inactive, select Inactive.
Figure 2. Source diagnostic cartridge in the Inactive position
-
On the bottom-left side of the Tune application, select the Diagnostics
icon.
The Diagnostics pane opens.
-
Select all of the following diagnostic tests:
-
Hardware Readbacks
-
Motor Movement Test
-
Cartridge Heater Test
-
Cartridge Sheath Gas Test
NOTE
You will perform the cartridge spray current test later in the installation process using an infusion cartridge.
Figure 3. Select the diagnostic tests -
-
Select Start.
The diagnostic tests are performed in sequential order.
-
During the Motor Movement test, use the Camera pane to confirm the following:
-
Motors move smoothly and quietly.
-
At the end of the Motor Movement Max test, the cartridge is positioned to the left of and above the inlet. The cartridge should also be close to the inlet on the z axis.
Figure 4. Proper cartridge position after the Motor Movement Max test NOTE
If you want additional time to visually confirm the cartridge position shown in the figure above, you can repeat the Motor Movement Max test after the diagnostic tests are complete. Afterward, return the cartridge to the Inactive position.
NOTE
If you observe any issues with the motor movement or cartridge position, contact your Thermo Fisher service engineer.
-
-
When the diagnostic tests are complete, view the diagnostic report to confirm that all of the tests received passing results.
NOTE
If you receive a failed result on any of these tests, see the Troubleshooting failed diagnostic tests section of the OptiSpray user guide, or contact your Thermo Fisher service engineer.