View safety notices
Make sure you understand the special notices, symbols, and caution labels in this guide. Most of the special notices and cautions appear in boxes, and those pertaining to safety also have corresponding symbols. Some symbols are also marked on the instrument itself and can appear in color or in black and white.
Notice, symbol, or label
Meaning
IMPORTANT
Highlights information necessary to prevent damage to software, loss of data, or invalid test results; or might contain information that is critical for optimal performance of the product.
Note
Highlights information of general interest.
Tip
Highlights helpful information that can make a task easier.
GUID-CE6E4EC1-84ED-42CC-BC1A-1D45FF6CEAF2-print.png
Caution: Read the cautionary information associated with this task.
GUID-72A4DC1F-AA52-4BEB-A725-1EBA0F8B4AC9-print.png
Chemical hazard: Observe safe laboratory practices and procedures when handling chemicals. Only work with volatile chemicals under a fume or exhaust hood. Wear gloves and other protective equipment, as appropriate, when handling toxic, carcinogenic, mutagenic, corrosive, or irritant chemicals. Use approved containers and proper procedures to dispose of waste oil and when handling wetted parts of the instrument.
GUID-1958D869-D640-4490-9CD9-9065A72738EA-print.png
Hot surface: Before touching the instrument, give the heated components time to cool.
GUID-A8380102-AC54-4D72-BED9-418B4C3971C6-print.png
Pinch point: Keep hands away from the specified areas.
GUID-E949EE62-182A-4917-B894-0229E857C61F-print.png
Electric shock: This instrument uses voltages that can cause electric shock and/or personal injury. Before servicing, shut down the instrument and disconnect it from line power. While operating the instrument, keep covers on.
GUID-10A71BDB-CD2C-4A81-AE94-D75931E3F8FF-print.png
Eye injury: Eye injury can occur from splattered chemicals, airborne particles, or sharp objects. Wear safety glasses when handling chemicals or servicing the instrument.
GUID-9BACC654-CFEF-4267-AFBA-9C6AE8EAEC02-print.png
Sharp object: Use caution when handling sharp elements (e.g. needles, blades) to avoid injury.
GUID-934976A1-2656-4B6B-92E9-E5F9AB38AC14-print.png
Trip obstacle: Be aware of cords, hoses, or other objects located on the floor.
GUID-61C57280-EC36-4D46-9903-25C9BD2B70E7-print.png
Crush injury: Be aware of falling objects that could cause foot injuries.
GUID-22047938-6E47-4B51-A569-933ED575CF4B-print.png
GUID-38031366-DB9D-427A-B6D0-3E5BB0105D61-print.png
Magnetic field hazard: This instrument generates magnetic fields that can interfere with cardiac implants. If you have a pacemaker or implantable cardioverter-defibrillator (ICD), maintain a minimum distance of 6 inches (15 centimeters) between the instrument and your implanted device. Closer proximity might cause device interference or malfunction.