The following table describes the information in the run queue table on the Queue page.
Column | Description |
|---|---|
Record Number | Displays the sequential number of the experiment in the run queue |
Experiment Name | Displays the name of the experiment |
Status | Displays the real-time status of the job:
|
Experiment Type | Displays the experiment workflow:
|
Method Name | Displays the processing method assigned to the experiment |
Method Type | (For Peptide Mapping Analysis experiments only) Displays the processing method subtype:
|
Sequence Names | (If applicable) Displays the name of the sequence(s) assigned to the experiment |
Raw File Names | Displays the name(s) of the raw data file(s) loaded for the experiment |
Source Spectra Method | (For Intact Mass Analysis experiments only) Displays the method used to detect the source spectra for deconvolution:
|
Deconvolution Algorithm | (For Intact Mass Analysis experiments only) Displays the algorithm used for deconvolution:
|
Number of Chromatographic Peaks | (For Intact Mass Analysis experiments using Auto Peak Detection or Average Over Selected Retention Time methods only) Displays the number of chromatographic peaks detected in the spectrum |
Number of Components Detected | (For Intact Mass Analysis experiments only) Displays the number of components detected after deconvolving the averaged spectrum of all chromatographic peaks. |
Submit Time | Displays the date and time the experiment was submitted to the run queue. |
Start Time | Displays the date and time that the application started processing the experiment. |
Completion Time | Displays the date and time that the application completed processing the experiment. |
Total Processing Time (min) | Displays the total time used to process the experiment. |