Right-click the Results table on the Coverage page to open a shortcut menu with the commands listed in the following table.
Command | Description |
---|---|
Show Oligonucleotide Coverage Map Parameters | Opens the Coverage Map Options dialog box to modify the oligonucleotide recovery parameters and the number of nucleotides per row displayed on the Oligonucleotide Sequence Coverage Map. See Modify Oligonucleotide Sequence Coverage Map parameters. |
Export | Exports all data (at the oligonucleotide- and raw data file-levels) in the Coverage Results table to an Excel workbook (.xlsx). |