Right-clicking the MS2 Spectra pane on the Process and Review page opens a shortcut menu with the commands listed in the following table.
Table MS2 Spectra pane shortcut menu
Command
Description
Reset Scale
Restores the original scale to the MS2 Spectra pane.
Copy
Copies the image viewable in the MS2 Spectra pane, including labeling and shading.
Predict Oligo MS/MS (Kinetic Model)
Uses the Kinetic model to generate the predicted spectrum in the MS2 Spectra pane.
Delete Predicted MS/MS Spectrum
Deletes the predicted spectrum in the MS2 Spectra pane.
To redisplay the predicted spectrum, you can either reselect the component row in the Results table or right-click in the MS2 Spectra pane and then select the Predict Oligo MS/MS (Kinetic Model) command from the shortcut menu.