The Trend Ratio pane on the Process and Review page displays bar plots representing the ratios between various experiment conditions and the reference condition for a selected component. The Trend Ratio pane is visible only for experiments with multiple raw data files.
Procedure
  1. Open the results of your experiment from the Queue page or from the Load Results page.
  2. See Open results.
  3. The Process and Review page opens by default and displays the results of your Oligonucleotide Analysis experiment, including the Trend Ratio plots for experiments with multiple raw data files.
  4. On the left side of the Process and Review page, select the Trend Ratio tab.
  5. Select the row for a component (or a specific raw data file under a component) in the Results table.
  6. The Trend Ratio plot shows the ratio (Condition/Reference Condition) for the selected component, with each condition depicted in a different color.
  7. The ratio values in the plot come from the associated columns in the Results table.
  8. The order of the conditions underneath the bars in the plot is the same order as the conditions entered on the Oligonucleotide Analysis page.
Figure Trend Ratio pane
Trend Ratio pane