The Save Method page contains two sections: Method Name/Notes (editable) and a read-only section summarizing experimental data, oligonucleotide sequence, ribonuclease, modifications, and processing parameters.
Figure Save Method page navigation
12345
No.DescriptionNo.Description
1
Parameters tab (processing method editing wizard)
4
Experiment information report area
2
Subpage navigation bar
5
Finish button
3
Method Name and Notes area
Once you have entered a method name, select the Finish button to save the method and submit the experiment for processing.
IMPORTANT
You cannot overwrite a default processing method. You must use a new name to save changes made to a default method.