Right-clicking the Components table on the Modification Summary page opens a shortcut menu with the commands listed in the following table.
Command | Description |
|---|---|
Export All Components | Exports all data in the Components table to an Excel workbook (.xlsx). |
Export Checked Components | Exports data only for the selected components in the Components table to an Excel workbook (.xlsx). |
Add to Report | (Ardia Platform mode only) Sources all or selected results for the report. |