Insert a report variable - Insert a report variable - Oligonucleotide Analysis User Guide - BioPharma Finder Software - Oligonucleotide Analysis User Guide - Insert a report variable - Report: Design page navigation - Reporting - BioPharma Finder Software - BioPharma Finder Software - topic
Oligonucleotide Analysis User Guide
- Product Tree
- Desktop Software > BioPharma Finder Software
- Ardia Platform > Software > BioPharma Finder Software
- Document Type
- User Guide
- Software version
- 5.5
- You have loaded experiment results from the Ardia Platform data repository.
- You are viewing the Design page on the Report tab.
- On the Design page, select a cell in the cell grid on the report sheet where you want to insert a report variable.
- Under the Insert tab, select Report Variable and then select General Information.
- The Insert Report Variable window opens, displaying a dropdown list of report variables.
Figure Insert Report Variable window- In the Insert Report Variable window, select a report variable from the dropdown list, and then select OK.
- The selected report variable is inserted in the report.