Upload default report templates to the Ardia Platform data repository - Upload default report templates to the Ardia Platform data repository - Oligonucleotide Analysis User Guide - BioPharma Finder Software - Oligonucleotide Analysis User Guide - Add default BioPharma Finder report templates to the Ardia Platform data repository - Upload default report templates to the Ardia Platform data repository - Reporting - BioPharma Finder Software - BioPharma Finder Software - topic
Oligonucleotide Analysis User Guide
- Product Tree
- Desktop Software > BioPharma Finder Software
- Ardia Platform > Software > BioPharma Finder Software
- Document Type
- User Guide
- Software version
- 5.5
- Start the BioPharma Finder application in Ardia Platform mode.
- On the BioPharma Finder Home page, in the Experiment Types pane, select the appropriate workflow link.
- The corresponding workflow page opens.
- On the navigation bar, select the Load Results tab.
- The Ardia Platform data repository folder tree opens.
- Open any experiment from the Ardia Platform data repository to enable the BioPharma Finder Report pages.
- On the Report page, select the Design button to view the Design subpage (if necessary).
- On the Design subpage, select Upload on the Utilities menu.
- The Upload Report Template window opens.
- At the top of the Upload Report Template window, select the Select Target Location and Name button.
- The Choose Ardia Repository Folder opens.
- In the Choose Ardia Repository Folder dialog box, navigate to the "ReportTemplates" folder in the folder tree and then select it.
- Enter a name for the report template in the Name box, and then select Save.
- The Upload Report template window opens again, displaying the chosen file path and name.
- At the bottom of the Upload Report Template window, select the Select File to Upload button.
- In the Open File window, navigate to the location on your computer where you saved the downloaded default report templates.
- Select the report template that you want to upload and then select Open.
- Select Upload.
- The default report template is added under the specified name in the "ReportTemplates" folder in the Ardia Platform data repository.
- Repeat these steps to add additional default templates to the folder as necessary.