In the Oligonucleotide Sequence Editor, you can save an oligonucleotide sequence as new sequence or overwrite an existing sequence.
Save a new sequence
Prerequisites
- You are editing an oligonucleotide sequence in the Oligonucleotide Sequence Editor.
Procedure
- In the Oligonucleotide Sequence Editor, select Save As New on the command bar.
- The Save As New dialog box opens.
Figure Save As New dialog box - In the New Sequence Name box, type a name for the new protein sequence.
- (Optional) In the Description box, type a description of the new protein sequence.
- Select OK.
- The new sequence is added to the Sequence Manager table.
Overwrite an existing sequence
Prerequisites
- You are editing an oligonucleotide sequence in the Oligonucleotide Sequence Editor.
Procedure
- In the Oligonucleotide Sequence Editor, select Save on the command bar.
- If the sequence is new, the sequence is added to the Sequence Manager table using the specified name/description.
- If the sequence exists, an overwrite warning prompt appears.
- Select Yes to overwrite the existing sequence with the new sequence.
- The sequence is updated in the Sequence Manager table.