The following table describes the types of information on the Coverage page.
Table Coverage page parameters
Parameter
Description
Coverage Results table
At the top level, displays the identified and unknown oligonucleotides, along with their designated color-coding flags used in the Coverage chromatogram. At the next level, the table displays the raw data files. At the lowest level, it displays the component information.
Coverage Chromatogram pane
Displays the chromatogram related to the row that you select in the Coverage Results table.
Relative Intensity (y axis)
Displays the ratio of each peak intensity relative to the maximum intensity in the spectrum.
RT (min) (x axis)
Displays the retention time (RT) of the spectrum, which is the time after injection at which a compound elutes. Retention time can also refer to the total time that the compound is retained on the chromatographic column.
Oligonucleotide Coverage Map pane
Displays the Oligonucleotide Sequence Coverage Map including the oligonucleotides, sequence coverage information, and the color-coded nucleotides.