The following table describes the commands on the Process and Review page.
Table Commands on the Process and Review page
Command
Description
Process
Reprocesses the Oligonucleotide Analysis experiment and deconvolves the source spectra with any changes made in the Real-Time Optimization pane on the Process and Review page. To activate the Process button, you must modify the experiment parameters.
See Use real-time optimization (RTO) to reprocess results.
Select the Process button to open a dialog box where you can enter a new experiment/method name or retain the same name to overwrite previously saved results/parameters in the current experiment with the new data.
Use only alphanumeric, space, underscore "_", and period "." characters, up to 50 maximum, in the experiment and method names.
You cannot overwrite an experiment that is currently submitted to the run queue or is actively processing or canceling. You also cannot overwrite a default method.
Manual Integrate
Performs a manual integration of a user-defined peak area on the selected ion chromatogram (SIC) for a component.
Save Results As
Saves the latest results in the database.
Select this button to open a dialog box where you can enter a new experiment name or retain the same name to overwrite previously saved results in the current experiment with the new results. You can also enter a description for the experiment.
This button is inactive if you modified any processing method parameter for real-time optimization. In this case, select Process to reprocess the experiment and reactivate this button.
Use only alphanumeric, space, underscore "_", and period "." characters, up to 50 maximum, in the experiment name.
You cannot overwrite an experiment that is currently submitted to the run queue or is actively processing or canceling.