The BioPharma Finder application uses the kinetic model to predict oligonucleotide MS/MS spectra. In the MS2 Spectra pane on the Process and Review page, you can change the parameters to regenerate the predicted spectrum using the model.
Prerequisites
- For an Oligonucleotide Analysis experiment, you have processed your experiment and are viewing the results on the Process and Review page.
Procedure
- On the Process and Review page, select the MS2 Spectra subtab.
- Select a row in the Results table that displays "MS2" in the ID Type column.
- Right-click the MS2 Spectra pane and select Predict Oligo MS/MS (Kinetic Model).
- The application opens the Predict Oligo MS/MS (Kinetic Model) dialog box.
Figure Predict Oligo MS/MS (Kinetic Model) dialog box - In the Sequence box, type the three-letter codes for a nucleotide sequence.
- Enter values for the following parameters:
- To set the charge state, the isolation width used for fragmentation, the resolution at m/z = 400, and the activation time (ms), enter a value in their boxes.
- To define a fragmentation method, select from the available options in the list.
- To choose an instrument, select an instrument name from the list.
- To define the collision energy or reagent target, enter a value in the box.
- Specify normalized collision energy as a percentage value (%) or specify the reagent ion target value for the ETD fragmentation method.
- Select OK to view the predicted MS/MS spectrum.
- The application displays the changes in the predicted spectrum in the MS2 Spectra pane.
- See View the predicted and experimental MS/MS spectra.