Default and custom processing methods
The BioPharma Finder application provides the Basic Default Method processing method optimized for Oligonucleotide Analysis experiments.
You can edit the processing parameters in the default method and save the customized method to apply it to your experiment.
| No. | Description | No. | Description |
|---|---|---|---|
1 | Select a processing method | 3 | Open the processing method editing wizard to edit parameters |
2 | Start processing (without editing parameters) |
On the Oligonucleotide Analysis page, in the Processing Method area, select a method and then select the Edit Method button to open the processing method editing wizard.
Edit a processing method
Prerequisites
- You are viewing the Oligonucleotide Analysis page.
- On the Oligonucleotide Analysis page, you have entered an experiment name, loaded the raw data files and entered the conditions (if applicable), and selected an oligonucleotide sequence (optional).
Procedure
- On the Oligonucleotide Analysis page, in the Processing Method area, select the checkbox for the processing method you want to edit for your experiment.
- Select the Edit Method button.
- The processing method editing wizard opens. By default, the Component Detection subpage is displayed.
Figure Processing method editing wizard subpage navigation No. Description No. Description 1
Edit Component Detection parameters
3
Enable/edit theoretical digestion and alias naming parameters
2
Edit Identification parameters
4
Save changes as a new method
- Navigate through the subpages of the wizard and edit the parameters as appropriate:
- On each subpage, select the Next button to progress to the next subpage of the wizard.
- Or, on the wizard navigation bar, select a subpage tab to open that subpage.
- The table below describes the parameters in each subpage of the processing method editing wizard.
- For more information on a subpage, select the subpage link in the table.
IMPORTANT
If you modify any of the parameters in a processing method, you must save the edited method using a new name to preserve your modifications.
Wizard subpage | Summary of parameters |
|---|---|
| |
| |
| |
|
- On the last page, the Save Method page, save the processing method.
- The application saves the processing method and the method appears in the list in the Processing Method area on the Oligonucleotide Analysis page. You can then select the method to process your current and future experiments.