For information about how to install an instrument driver package, refer to the installation instructions provided with the relevant MS driver.
For information about specific versions of the instrument drivers supported by Chromeleon, refer to the List of Supported Instruments. The document can be accessed through Windows Start > Thermo Chromeleon 7 > Documents or can be found in the Documents folder on the Chromeleon installation medium.
Control of Thermo Scientific mass spectrometers is provided by separately installed drivers. Customers with support contracts might access the following download locations for the current drivers, as well as important information about model and version compatibility:
If you do not have access to either of the above locations, contact your local Thermo Scientific representative.