Use Xcalibur or TraceFinder to control your LCMS or GCMS - Chromeleon CDS Software - Chromeleon 7.3.2 MUc MS Software Installation Guide - Installation on a PC with Xcalibur/TraceFinder installed - Use Xcalibur or TraceFinder to control your LCMS or GCMS - Xcalibur, TraceFinder and Chromeleon - Chromeleon CDS Software - Chromeleon CDS Software
MS Software Installation Guide
- Product Tree
- Ardia Platform > Software > Chromeleon CDS Software
- Desktop Software > Chromeleon CDS Software
- Document Type
- Installation Guide
- Software version
- 7.3.2 MUc
- Stop the Chromeleon Instrument Controller.
- Right-click the Chromeleon tray icon in the notification area of the Windows task bar.
- Select Stop Chromeleon Instrument Controller.
- Add the LC, GC, and MS hardware drivers to the Foundation Instrument Configuration.