The NIST Mass Spectral Search Program is not included in the Chromeleon setup. If you have purchased a full copy of any of the NIST MS libraries, the library installation also installs the NIST Mass Spectral Search and AMDIS software.
If required, for example for the MS Library search, you might install a demo version of the program manually from the installation medium (in the Tools folder on the Chromeleon installation medium).
NOTE
An upgrade from an earlier Chromeleon version to Chromeleon 7.2 SR5 or later might remove the NIST Mass Spectral Search program (if installed) from your computer.
  • Reinstall the NIST Mass Spectral Search program after the Chromeleon upgrade, if necessary.