Start the Instrument Configuration Manager - Chromeleon CDS Software - Chromeleon 7.3.2 MUc MS Software Installation Guide - Start the Instrument Configuration Manager - Install and configure MS software components - Configure MS instruments in Chromeleon - Chromeleon CDS Software - Chromeleon CDS Software
MS Software Installation Guide
- Product Tree
- Ardia Platform > Software > Chromeleon CDS Software
- Desktop Software > Chromeleon CDS Software
- Document Type
- Installation Guide
- Software version
- 7.3.2 MUc
- In the Chromeleon Services Manager, select Configure Instruments (only available if the Instrument Controller Service is running).
– OR –
- Select Start > Programs > Thermo Chromeleon 7 > Instrument Configuration Manager.