You can update settings like filters for all XICs simultaneously with the following steps.
Procedure
  1. Open the MS Component Table in the Processing Method pane.
  2. Just above the table header, select Show Properties.
  3. The dialog box with the properties of the currently selected component opens.
  4. In the dialog box, select Extracted Ion Chromatograms.
  5. In the XICs area, select Update.
  6. The Update XIC Settings dialog box opens.
  7. Select For All Components.
  8. NOTE
    Other options, like smoothing, can be applied per component. In that case you can select For Selected Component and specify one or multiple components. Note that as any change per component, this can affect all injections that use the same processing method.
  9. Clear the Calculation Ion Ratios option.
  10. This option is selected mainly for small molecules.
  11. Select Update Filters for All Ions.
  12. Select Use This Filter and then select an appropriate full-MS filter from the drop-down list.