After you registered with the Ardia Platform through Chromeleon CDS software, you can open Chromeleon CDS software in Ardia Platform mode.
Procedure
- If not already enabled, open the Chromeleon Administration Console, and select Global Policies > User Mode > Enable Ardia Integration.
- If the Ardia Platform integration is successful, a connection URL is displayed under the Enable Ardia Integration check box.
TIP
Select Enable Single Sign-On to allow the user to start the Chromeleon CDS software without having to enter their credentials if the Windows session already has an active Single Sign-On session.
- Open the Chromeleon CDS software.
- If you registered with the Ardia Platform and enabled connection to the Ardia Platform, the Chromeleon CDS software opens in Ardia Platform mode and prompts you to sign in.
- Sign in.