When you have defined all the sheets of the report template, also consider the following:
  • Set up headers and footers for the report pages, for example, to include template sheet names page and numbers.
  • Set print areas so that the report tables fit into the width of a single printed page.
  • From the Print dialog, set up the correct conditions for when to print which report sheet. For example, only print the Sequence overview for the first injection, the SST report for injections of the Check Standard type, the modification reports for injections of Unknown type, the new peak detection report for injections that contain the word Spiked in their name, and so on.