As a crucial piece of information to document the performance of your system and to support the validity of your data, insert a report sheet that reports the results of the test cases you have defined for the SST.
You can insert a predefined table template by selecting Test Cases Table from the Results group. Consider editing the predefined template, for example:
- Some test cases might have two reference values, so you might want to add a column that reports the values of variable
sst.reference_value("2nd")
. - You might also want to have a column that reports the evaluation result value
sstResult.eval_result
that the test case result was based on.
NOTE
The sheet of the report template can be reused to report data from different injections. The values in the table change automatically when you select another injection in the navigation pane. Include the injection name filed above the SST table, so that it is always clear what data is listed in the table.