To view the results of the calculations that Chromeleon CDS software performed to evaluate your test cases, as well as the overall SST results, display the Interactive Results pane and insert the System Suitability IRC table from the Processing Method group. Refer to the Chromeleon CDS Software User Help for details related to viewing interactive result tables.
TIP
The overall result of the SST is also indicated by the color of the injection icon, either in the navigation pane in the Chromatography Studio, or in the injection list in the Console.
To be able to switch quickly between the view of the Chromatography Studio dedicated to check standards and the view dedicated to inspecting sample injections, save the current view that includes the interactive result table as a separate view settings file.