After an injection has been processed by the NTMS processing, the navigation pane displays an icon that indicates the overall injection scoring. An injection is scored as In Spec if there are no new peaks listed after the filters have been applied, otherwise the injection is scored as Not In Spec.
Note that injection scoring displayed in the navigation pane of the NTMS Processing category depends on the current component result filters, and therefore on the current view settings. For scoring injections in reports, see Reporting PQA monitoring results in the Chromeleon CDS software.