Isotopes of native and corresponding deamidated peptides have very similar masses. In this case, use the option to specify mass tolerance per component and decrease its value for the peptide and its deamidated form from the global default mass tolerance to a lower value. In addition, you can define the retention time of the deamidated peptide relative to the retention time of the native peptide.