You can filter and search within the Results table to refine the displayed results and to view specific PQAs.
Prerequisites
  1. You have opened your peptide mapping analysis results and are viewing the Process and Review page.
Procedure
  1. In the Results table, do the following to filter the table using the Identification column:
  2. Select the down arrow in the column header.
  3. Select (NonBlanks) from the dropdown list.
  4. The Results table displays only the results of components that are identified in the analysis.
  5. NOTE
    Identified components are components whose deconvoluted masses match the theoretical masses of peptides and their modifications derived from the protein sequence assigned to the experiment.
  6. (Optional) Filter the Results table using other columns or by using a saved custom filter file.
  7. TIP
    For more information on using custom filters, refer to the BioPharma Finder Software User Guide.
  8. (Optional) Search for specific results (PQAs) by entering key words into any of the column header boxes.
  9. For example, to find results associated with the peptide DTLMISR, enter "DTLMISR" in the Peptide Sequence column header box. Or, to view all oxidized components in the analysis, enter "Oxidation" in the Modification column header box.